Human recombinant IL-17A
ENZOENZPRT1880025
:
- Pk:25 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Protein synonyms:CTLA8|Interleukin-17|Cytotoxic T-lymphocyte-associated serine esterase 8
- Protein/peptide name:IL-17A
- Purity:>98% (SDS and HPLC)
- Molecular weight:~31,3 kDa
- Formulation:Lyophilized with no additives.
- Purification:Sterile filtered through a 0,2 micron filter.
Produced in E. coli.