Order Entry
Spain
ContactUsLinkComponent
Human recombinant ACE1
Human recombinant ACE1
Catalog # ENZOENZPRT1560010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant ACE1
Catalog # ENZOENZPRT1560010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein synonyms:
    CD143|MVCD3|DCP|DCP1|MGC26566|Angiotensin-converting enzyme 1|Peptidyl-Dipetidase A
  • Protein/peptide name:
    ACE1
  • Purity:
    >95% (SDS-PAGE)
  • Molecular weight:
    ~170 kDa
  • Formulation:
    Liquid. Contains sodium chloride, ZnCl2, TRIS and glycerol

Specifications

About this item

Produced in NSO cells.