Human recombinant stem cell factor
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Produced in E. coli. Non-glycosylated protein, containing 165 amino acids.
- High purity
- Carrier-free
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Biological activity:The activity is determined by the dose-dependent stimulation of human TF-1 cells which is typically observed at a concentration of 1 - 5 ng/ml.
- Protein synonyms:c-Kit ligand|SCF|CD117|Steel factor|KL|MGF|c-kit
- Protein/peptide name:Stem cell factor
- Purity:>98% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
- Molecular weight:~18,4kDa
- Formulation:Lyophilized from 10 mM acetic acid (AcOH) at a concentration of 1 mg/ml. Sterile filtered.
Specifications
Frequently Bought Together