Human recombinant UBCH10
Catalog # ENZOBMLUW09600100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Protein/peptide name:UBCH10
- Purity:>95% (SDS-PAGE)
- Molecular weight:~19,7 kDa
- Concentration:0,5 mg/ml
- Formulation:Liquid. In PBS.
Specifications
About this item
Produced in E. coli.