Human recombinant Urm1 (from E. coli), HIS tag
Catalog # ENZOBMLUW95300100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:URM1
Specifications
About this item
Produced in E. coli.