Order Entry
Spain
ContactUsLinkComponent
Human recombinant MMP20 (catalytic domain) (from E. coli)
Human recombinant MMP20 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE5400010
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant MMP20 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE5400010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    MMP20 (catalytic domain)

Specifications

About this item

Produced in E. coli. Active recombinant matrix metalloproteinase-20 (MMP-20, enamelysin) cloned from human cDNA. The enzyme consists of residues Tyr108-Phe276 (NM_004771), which comprises the catalytic domain of human MMP-20, with a C-terminal purification tag. This represents a naturally-occurring active form of MMP-20 which lacks the C-terminal hemopexin domain.

MMP-20 functions in tooth development and is implicated in cancer and oral pathology.