Human recombinant PDE10A (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE5250020
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:PDE10A (catalytic domain)
Specifications
About this item
Produced in E. coli. Active catalytic domain of phosphodiesterase 10A.