Human recombinant Cyclophilin A (from E. coli)
ENZOBMLSE1050100
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:Cyclophilin A
- Purity:>95% (SDS-PAGE)
- Formulation:Liquid. In 20 mM Tris-HCl, pH 7,8.
Produced in E. coli.