Order Entry
Spain
ContactUsLinkComponent
Human native ceruloplasmin (from plasma)
Human native ceruloplasmin (from plasma)
Catalog # ENZOALX200089M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human native ceruloplasmin (from plasma)
Catalog # ENZOALX200089M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Native
  • Source:
    Plasma
  • Species:
    Human
  • Seq:
    02
  • Reconstitution instructions:
    Reconstitute with distilled water.
  • Protein/peptide name:
    Ceruloplasmin
  • Purity:
    >95% (SDS-PAGE)
  • Formulation:
    Lyophilised from 50 mM potassium phosphate, pH 6,8, containing 100 mM potassium chloride, 20 mM ε-aminocaproic acid and 5 mM EDTA.
  • Purification:
    Plasma has been tested negative for HBsAg and for antibodies to HIV and HCV

Specifications

About this item

Serum copper transport protein

Isolated from human plasma.

Serum copper transport and iron-oxidizing protein. Expressed in plasma at concentrations of 1-5 µM. Contains six copper centers and is known to oxidize amines in a process coupled to the reduction of molecular oxygen. Has ferroxidase activity that is responsible for the oxidation of ferrous iron to its ferric form, which is necessary for efficient iron efflux from the cell (e.g. during hypoxia). Has previously been considered a target for nitric oxide (NO) and an inhibitor of endothelial nitric oxide synthase (eNOS/NOS III). Catalyzes S-nitrosothiol formation in cell culture media. Is a NO oxidase and nitrite synthase that determines endocrine NO homeostasis.