Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Angiotensin I
Angiotensin I
Angiotensin I
Catalog # ENZOALX151037M005
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Angiotensin I
Catalog # ENZOALX151037M005
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
5 mg
Seq:
02
Protein/peptide name:
Angiotensin I
Purity:
>97% (HPLC)
Specifications
About this item
Product Details
Product Details