Human native HSP90 (from HeLa cells)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Produced in HeLa cells.
Specifications
- Protein/peptide type:Native
- Source:HeLa cells
- Species:Human
- Protein/peptide name:HSP90
- Purity:-
- Formulation:-
- Purification:-
Specifications
Frequently Bought Together