Order Entry
Spain
ContactUsLinkComponent
Rat recombinant HSP70/HSP72 (from E.coli)
Rat recombinant HSP70/HSP72 (from E.coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
Rat recombinant HSP70/HSP72 (from E.coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Protein/peptide name:
    HSP70/HSP72

Specifications

Frequently Bought Together