Order Entry
Spain
ContactUsLinkComponent
Rat recombinant HSP60 (from E. coli)
Rat recombinant HSP60 (from E. coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
Rat recombinant HSP60 (from E. coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Protein/peptide name:
    HSP60
  • Purity:
  • Formulation:
  • Purification:

Specifications

Frequently Bought Together