Order Entry
Spain
ContactUsLinkComponent
Mouse recombinant HSP70-A1 (from E. coli)
Mouse recombinant HSP70-A1 (from E. coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
Mouse recombinant HSP70-A1 (from E. coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Protein/peptide name:
    HSP70-A1
  • Purity:
    >85% (SDS-PAGE; Western blot)
  • Formulation:
    Liquid, in Dulbecco’s PBS, pH 7,5, containing 5,0 mM glutathione
  • Purification:
    Purified by multi-step chromatography

Specifications

Frequently Bought Together