Order Entry
Spain
ContactUsLinkComponent
Anti-MAP2K1 + MAP2K2 Mouse Monoclonal Antibody [clone: 7E10]
Anti-MAP2K1 + MAP2K2 Mouse Monoclonal Antibody [clone: 7E10]
Catalog # ENZOADI905652100
Supplier:  Enzo Life Sciences
Anti-MAP2K1 + MAP2K2 Mouse Monoclonal Antibody [clone: 7E10]
Catalog # ENZOADI905652100
Supplier:  Enzo Life Sciences

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated Protein Kinase Kinase 1 And 2
  • Antigen symbol:
    MAP2K1 + MAP2K2
  • Clonality:
    Monoclonal
  • Clone:
    7E10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Dog,
    Mouse
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Lyophilized
  • Antigen synonyms:
    PRKMK2|MAPKK 2|MAPKK 1|MAP kinase kinase 2|MAPK/ERK kinase 2|Map2k2|dual specificity mitogen-activated protein kinase kinase 1|MAP2K1|MEK 1|ERK activator kinase 2|ERK activator kinase 1|MEK2|MKK2|Dual specificity mitogen-activated protein kinase kinase 2|MAP kinase kinase 1|MAPK/ERK kinase 1|mitogen-activated kinase 1
  • Modification:
    phospho-Ser218, phospho-222
  • Storage buffer:
    PBS containing 0,09% sodium azide, PEG, and sucrose.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Synthetic peptide corresponding to a portion of MEK1 phosphorylated at Ser218/222and MEK2 phosphorylated at Ser222/226 , conjugated to KLH.
  • Purification:
    purified by thiophilic adsorption and size exclusion chromatography
  • Pk:
    100 µG

Specifications

About this item

MEK is a dual specificity kinase capable of phosphorylating both tyrosine and threonine residues. The MEK family, which is also known as MAP kinase kinase, phosphorylates MAP kinases on the conserved T-X-Y motif; phosphorylation of MAPK by MEK results in an increase in MAPK activity. MEK is involved in a diverse array of cellular processes such as stress-activated response, apoptosis, cytokine-induced cell proliferation, and DNA recombination during meiosis.

Type: Primary
Antigen: MAP2K1 + MAP2K2
Clonality: Monoclonal
Clone: 7E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse