Distillation condenser glass
Catalog # GERH1000058
Supplier: GERHARDT
Specifications
- Description:Distillation condenser glass DK8 with long outlet pipe
- Pk:1
Specifications
About this item
This distillation condenser glass has a connecting piece for the ventilation valve.