Sealing ring
Sealing ring for filling valve, FKM, Black
Catalog # BRND6694
Supplier: Brand
Jump to:
Product Details & Documents
This sealing ring is made from FKM and is only suitable for autoclaving applications.
Specifications
- Colour:Black
- Description:Sealing ring for filling valve, FKM
- Cat. No.:BRND6694
- Environmentally Preferable:Sustainable Packaging