Dutasteride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:164656-23-9
:
Formula:C₂₇H₃₀F₆N₂O₂
MDL Number:MFCD00937869
Melting Pt:245 °C
:528.54 g/mol
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- HPLC:≥98.5 %
Specification Test Results
Frequently Bought Together