Urine collection tubes, primary, with screw cap, WHEATON®
Supplier: DWK Life Sciences
Certificates
About this item
These WHEATON® round bottom tubes are manufactured from virgin PS. The tubes have a capacity of 11 ml and are fitted with a leakproof screw cap for ease of transportation and storage of the sample. The tubes are supplied with a collection cup for the patient’s comfort during sample collection, and for them to easily transfer the sample to the tube.
- Ideal for use with automated urine analysis machines
- Printed label for patient details
- Option of plain tube or pre-dosed with boric acid
- Removes the need for unnecessary decanting of sample in the laboratory
Delivery: Tubes are supplied with sample collection cup.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together