Order Entry
Spain
Orders LinkContactUsLinkComponent
Thrombin (biotinylated)
Thrombin (biotinylated)
Thrombin (biotinylated)
Certificates
2 Options Available Below
Product Details & Documents
Biotinylated Thrombin has covalently attached biotin for easy removal of the enzyme from cleavage reactions using immobilised streptavidin. This preparation is tested for activity using the same assay as for unmodified thrombin, and for >99% binding to streptavidin agarose.
Storage temperature: −20 °C.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 69672-500UN
  • 69672-1000UN
  • Enzyme name:
  • Thrombin (biotinylated)
  • Thrombin
Specifications
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 500 U/g
    Log in for availability
    Log in for availability
    1 * 1 C/U
    1 * 1 C/U - 2 970,00 €
    2 970,00 €
    1 * 1 C/U
       2 970,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    69672-500UN
    Enzyme name
    Thrombin (biotinylated)
  • 1.000 U/g
    Log in for availability
    Log in for availability
    1 * 1 C/U
    1 * 1 C/U - 4 810,00 €
    4 810,00 €
    1 * 1 C/U
       4 810,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    69672-1000UN
    Enzyme name
    Thrombin