Order Entry
Spain
Orders LinkContactUsLinkComponent
Etodolac, Reference Standards

Etodolac, Reference Standards

Supplier:   USP
Etodolac, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1268706.
  • 1268728.
  • 1268892.
  • Description:
  • Etodolac
  • Etodolac Related Compound A
  • Etodolac Peak Identication Mixture
  • Synonym(s):
  • (+/-)-8-Ethyl-1-methyl-1,3,4,9-tetrahydropyrano [3,4-b]-indole-1-acetic acid
  • CAS No.:
  • 41340-25-4
  • 109518-50-5
  • Formula:
  • C₁₇H₂₁NO₃
  • C₁₆H₁₉NO₃
  • UN No.:
  • 2811
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1268706.
    Etodolac
    Etodolac
    USP reference standard
    Vial
    400 mg
    Log in for availability
    Log in for availability
    1 * 400 mg
    1 * 400 mg - 412,00 €
    412,00 €
    1 * 400 mg
       412,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1268706.
    Description
    Etodolac
    Synonym(s)
    CAS No.
    41340-25-4
    Formula
    C₁₇H₂₁NO₃
    UN No.
    2811
  • 1268728.
    Etodolac Related Compound A
    Etodolac Related Compound A
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 240,00 €
    1 240,00 €
    1 * 25 mg
       1 240,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1268728.
    Description
    Etodolac Related Compound A
    Synonym(s)
    (+/-)-8-Ethyl-1-methyl-1,3,4,9-tetrahydropyrano [3,4-b]-indole-1-acetic acid
    CAS No.
    109518-50-5
    Formula
    C₁₆H₁₉NO₃
    UN No.
  • 1268892.
    Etodolac Peak Identication Mixture
    Etodolac Peak Identication Mixture
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 300,00 €
    1 300,00 €
    1 * 15 mg
       1 300,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1268892.
    Description
    Etodolac Peak Identication Mixture
    Synonym(s)
    CAS No.
    Formula
    UN No.
    2811
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.