Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, Sisomicin sulfate, Ph.Eur., RS
Organic reference standard, Sisomicin sulfate, Ph.Eur., RS
Catalog # EDQMS0660000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Sisomicin sulfate, Ph.Eur., RS
Catalog # EDQMS0660000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    60 mg
  • Standards:
    Reference standard
  • CAS No.:
    53179-09-2
  • Description:
    Sisomicin sulfate
  • Formula:
    C₃₈H₈₄N₁₀O₃₄S₅
  • Storage temperature:
    Ambient
  • Description:
    Sisomicin sulphate

Specifications

About this item

  • In Monograph 0331 and 1351
  • Partial solubility in water
  • Pharmaceutical applications