Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, Perindopril for stereochemical purity, Ph.Eur., RS
Organic reference standard, Perindopril for stereochemical purity, Ph.Eur., RS
Catálogo # EDQMY0000207
Supplier:  EDQM
CAS Number:  
Organic reference standard, Perindopril for stereochemical purity, Ph.Eur., RS
Catálogo # EDQMY0000207
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    40 mg
  • Standards:
    Reference standard
  • CAS No.:
    107133-36-8
  • Description:
    Perindopril for stereochemical purity
  • Formula:
    C₂₃H₄₃N₃O₅
  • Storage temperature:
    Ambient
  • Description:
    Perindopril for stereochemical purity

Specifications

About this item

  • In Monograph 2019
  • Soluble in ethanol (96%), dimethylsulfoxide (DMSO), and water
  • Pharmaceutical applications