Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, Naphazoline impurity A, Ph.Eur., RS
Organic reference standard, Naphazoline impurity A, Ph.Eur., RS
Catalog # EDQMN0200000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Naphazoline impurity A, Ph.Eur., RS
Catalog # EDQMN0200000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    15 mg
  • Standards:
    Reference standard
  • CAS No.:
    36321-43-4
  • Description:
    Naphazoline impurity A
  • Formula:
    C₁₄H₁₆N₂O
  • Storage temperature:
    Ambient
  • Description:
    Naphazoline impurity A

Specifications

About this item

  • In Monographs 0147 and 0730
  • Solubility in water: 100 - 1000 g/l