Order Entry
Spain
ContactUsLinkComponent
 
Organic reference standard, Endotoxin standard BRP, Ph.Eur., RS
  EDQME0150000
 :  EDQM
 :  
Organic reference standard, Endotoxin standard BRP, Ph.Eur., RS
  EDQME0150000
 :  EDQM
 :  

 

  • Pack type:
    Vial
  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    67924-63-4
  • Description:
    Endotoxin standard BRP
  • Storage temperature:
    -20°C with ice
  • Description:
    Endotoxin standard BRP

 

 

  • In Monographs 20614 and 20630
  • Freeze-dried material