Order Entry
Spain
ContactUsLinkComponent
Sodium thiosulphate 0.05 mol/l (0.05 N) in aqueous solution standard volumetric solution
Sodium thiosulphate 0.05 mol/l (0.05 N) in aqueous solution standard volumetric solution
  APPC182160.1211
 :  
Sodium thiosulphate 0.05 mol/l (0.05 N) in aqueous solution standard volumetric solution
  APPC182160.1211
 :  

 

  • Pk:
    1.000 mL
  • Seq:
    2865

 

 

  • Titer at 20 °C:
    0,999 - 1,001
  • Traceability:
    NIST
  • pH:
    9 - 10
  • Appearance:
    Clear colourless liquid
  • Solubility:
    Miscible in water
  • Storage:
    RT

 

 

 :
 :158.11 g/mol
 :7772-98-7
 :Na₂S₂O₃
 :MFCD00003499