Order Entry
Spain
ContactUsLinkComponent
Human recombinant IL-10 non-lytic (from NS1 cells)
Human recombinant IL-10 non-lytic (from NS1 cells)
Catalog # PRSI90-496
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL-10 non-lytic (from NS1 cells)
Catalog # PRSI90-496
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    NS1 cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Biological activity:
    Bioactivity demonstrated in cytokine synthesis inhibition assay, measuring inhibition of IL-6 production by PU5-1 cells.
  • Protein synonyms:
    Interleukin-10|Cytokine Sythesis Inhibitory Factor|CSIF
  • Protein/peptide name:
    IL-10 non-lytic
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    19-178
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.

Specifications

About this item

Interleukin-10 (IL-10) is a cytokine produced by activated Th2 cells, B cells, keratinocytes and monocytes/macrophages. In vitro murine and human IL-10 inhibits cytokine synthesis by Th1 cells, natural killer cells and monocytes/macrophages. Several studies have suggested the potential application of IL-10 as an anti-inflammatory agent in the treatment of septic shock and as an immunosuppressive agent in certain T cell mediated autoimmune diseases.

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.