Order Entry
Spain
ContactUsLinkComponent
Human recombinant Krueppel-like factor 6 (from E. coli)
Human recombinant Krueppel-like factor 6 (from E. coli)
Catalog # PRSI92-270
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Krueppel-like factor 6 (from E. coli)
Catalog # PRSI92-270
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    COPEB|Suppressor of Tumorigenicity 12 Protein|KLF6|Transcription Factor Zf9|BCD1|B-Cell-Derived Protein 1|GC-Rich Sites-Binding Factor GBF|Core Promoter Element-Binding Protein|CPBP|ST12|Proto-Oncogene BCD1|Krueppel-like Factor 6
  • Protein/peptide name:
    Krueppel-like factor 6
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    12.6 kD
  • Sequence:
    Met1-Ser109
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Krueppel-Like Factor 6 (KLF6) belongs to the krueppel C2H2-type zinc-finger protein family. KLF6 contains three C2H2-type zinc fingers and localises in the nucleus. KLF6 expression is highest in the placenta followed by spleen, thymus, prostate, testis, small intestinem and colon. However, it is weakly expressed in the pancreas, lung, liver, heart, and skeletal muscle. KLF6 functions as a transcriptional activator and could play a role in B-cell growth and development. Defects in KLF6 will result in gastric cancer and prostate cancer.

This recombinant protein can be used for biological assays. For research use only.