Order Entry
Spain
ContactUsLinkComponent
Human recombinant PLAU (from HEK293 cells)
Human recombinant PLAU (from HEK293 cells)
  PRSI96-630
 :  
undefined
Human recombinant PLAU (from HEK293 cells)
  PRSI96-630
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured by its ability to bind human PLAUR in a function ELISA.
  • Protein synonyms:
    URK|ATF|u-PA|Urokinase|PLAU|QPD|UPA|BDPLT5
  • Protein/peptide name:
    PLAU
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    45 kDa
  • Formulation:
    Lyophilized, 50 mM NaAc, 100 mM NaCl, pH5.0
  • Tested applications:
    Western Blot