West nile virus WNV envelope protein peptide (16 aa near N-terminus)
Catalog # PRSI3439P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:West nile virus
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa near N-terminus
- Protein/peptide name:WNV Envelope Protein Peptide (16 aa near N-terminus)
- Tested applications:Blocking
Specifications
About this item
West Nile virus peptide is used for blocking the activity of West Nile virus envelope antibody.