Order Entry
Spain
ContactUsLinkComponent
Avian influenza A (H5N1) neuraminidase Influenza A Peptide (15 aa near C-terminus)
Avian influenza A (H5N1) neuraminidase Influenza A Peptide (15 aa near C-terminus)
Catalog # PRSI3423P
Supplier:  ProSci Inc.
CAS Number:  
Avian influenza A (H5N1) neuraminidase Influenza A Peptide (15 aa near C-terminus)
Catalog # PRSI3423P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Avian influenza A (H5N1) neuraminidase
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    15 aa near C-terminus
  • Protein/peptide name:
    Influenza A Peptide (15 aa near C-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

The peptide is used for blocking the activity of the avian influenza neuraminidase antibody.