- Antigen name:Interleukin 1 Alpha
- Antigen symbol:IL1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Hematopoietin-1|IL-1 alpha|pro-interleukin-1-alpha|IL1A|preinterleukin 1 alpha|hematopoietin-1|interleukin-1 alpha
- Concentration:1 mg/mL
- Immunogen:1-271
- Size:50 µg
- Pk:50 µG
Interleukin 1, beta proprotein. IL-1beta is a member of the interleukin 1 cytokine family. This cytokine is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. This cytokine is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury, and thus induces apoptosis. The gene encoding this protein and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. It has been suggested that the polymorphism of these genes is associated with rheumatoid arthritis and Alzheimer's disease.
IL-1a antibody can be used for the detection of IL-1a by Western blot, may also work for IHC and ICC.
Antigen affinity-purified
Phosphate-Buffered Saline. No preservatives added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: IL1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat