Anti-KITLG Goat Polyclonal Antibody
PRSIXP-5265
- Antigen name:KIT ligand
- Antigen symbol:KITLG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Kit ligand|SCF|KL-1|Kitl|Stem cell factor precursor|SF|FPH2|Mast cell growth factor|MGF|SHEP7|KIT ligand
- Immunogen:Produced from sera of goats pre-immunized with highly pure (>98%) recombinant hSCF (human SCF).
- Size:100 µg
- Pk:100 µG
Anti-hSCF specific antibody was purified by affinity chromatography employing immobilized hSCF matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: KITLG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human