Specifications
- Antigen name:Hydroxysteroid (17-beta) Dehydrogenase 10
- Antigen symbol:HSD17B10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:MRX31|ERAB|17b-HSD10|MRX17|HADH2|MRPP2|HCD2|MRXS10|MHBD|SCHAD|CAMR|ABAD|SDR5C1|DUPXp11.22
- Concentration:1 mg/mL
- Immunogen:ERAB antibody was raised against a peptide sequence around aa.253~257 (L-D-G-A-I) derived from Human ERAB.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Functions in mitochondrial tRNA maturation. Part of mitochondrial ribonuclease P, an enzyme composed of MRPP1/RG9MTD1, MRPP2/HSD17B10 and MRPP3/KIAA0391, which cleaves tRNA molecules in their 5'-ends. By interacting with intracellular amyloid-beta, it may contribute to the neuronal dysfunction associated with Alzheimer disease (AD).
Western Blot: 1:500~1:1000
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: HSD17B10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat