Anti-ZNF703 Goat Polyclonal Antibody
Catalog # PRSI46-618
Supplier: ProSci Inc.
Specifications
- Antigen name:Zinc Finger Protein 703
- Antigen symbol:ZNF703
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Concentration:500 ug/mL
- Immunogen:ZNF703 antibody was raised against a 14 amino acid synthetic peptide near the internal region of ZNF703.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 28-30 kDa band in human breast lysates after 1 μg/mL antibody staining. Approximately 60 kDa band observed in human breast carcinoma lysates (calculated MW of 58.2 kDa according to NP_079345.1). Recommended concentration: 1-3 μg/mL.
ZNF703 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ZNF703 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZNF703
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human