Anti-Diphtheria toxin Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI35-158
Supplier: ProSci Inc.
Specifications
- Antigen name:Diphtheria toxin
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Goat
- Reactivity:Bacteria
- Western blot:Yes
- Form:Liquid
- Immunogen:Purified diphtheria toxoid
- Size:1 mL
- Pk:1 mL
Specifications
About this item
Purified IgG fraction of antiserum covalently coupled with high purity Isomer I of fluorescein isothiocyanate. Care is taken to insure complete removal of any free fluorescein from the final product.
Diphtheria toxin antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing BSA (10 mg/ml) as a stabilizer and 0.1% sodium azide preservative.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Diphtheria toxin
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Goat
Isotype:
Reactivity: Bacteria