Anti-IRX6 Rabbit Polyclonal Antibody
PRSI28-646
- Antigen name:Iroquois Homeobox Protein 6
- Antigen symbol:IRX6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:IRX7|IRXB3|IRX-3
- Immunogen:A synthetic peptide corresponding a region of human IRX6.
- Size:100 µg
- Pk:100 µG
IRX6 is one Iroquois (Irx) proteins comprise a family of homeodomain-containing transcription factors involved in patterning and regionalization of embryonic tissues in both vertebrates and invertebrates.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: IRX6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse