Order Entry
Spain
ContactUsLinkComponent
Anti-NEK6 Rabbit Polyclonal Antibody
Anti-NEK6 Rabbit Polyclonal Antibody
Catalog # PRSI26-344
Supplier:  ProSci Inc.
undefined
Anti-NEK6 Rabbit Polyclonal Antibody
Catalog # PRSI26-344
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    NIMA-Related Kinase 6
  • Antigen symbol:
    NEK6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    SID|SID6-1512|RP11-101K10.6
  • Immunogen:
    A synthetic peptide corresponding a region of human NEK6.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

The Aspergillus nidulans 'never in mitosis A' (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA.The Aspergillus nidulans 'never in mitosis A' (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NEK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat