Order Entry
Spain
ContactUsLinkComponent
Anti-INMT Rabbit Polyclonal Antibody
Anti-INMT Rabbit Polyclonal Antibody
  PRSI26-340
undefined
Anti-INMT Rabbit Polyclonal Antibody
  PRSI26-340

 

  • Antigen name:
    Indolethylamine N-methyltransferase
  • Antigen symbol:
    INMT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Immunogen:
    A synthetic peptide corresponding a region of human INMT.
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

N-methylation of endogenous and xenobiotic compounds is a major method by which they are degraded. This gene encodes an enzyme that N-methylates indoles such as tryptamine.N-methylation of endogenous and xenobiotic compounds is a major method by which they are degraded. This gene encodes an enzyme that N-methylates indoles such as tryptamine.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: INMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human