Order Entry
Spain
ContactUsLinkComponent
Anti-ADSSL1 Rabbit Polyclonal Antibody
Anti-ADSSL1 Rabbit Polyclonal Antibody
Catalog # PRSI26-312
Supplier:  ProSci Inc.
undefined
Anti-ADSSL1 Rabbit Polyclonal Antibody
Catalog # PRSI26-312
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Adenylosuccinate Synthase like 1
  • Antigen symbol:
    ADSSL1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Immunogen:
    A synthetic peptide corresponding a region of human ADSSL1.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

ADSSL1 is a muscle isozyme of adenylosuccinate synthase (EC 6.3.4.4), which catalyzes the initial reaction in the conversion of inosine monophosphate (IMP) to adenosine monophosphate (AMP).

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ADSSL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat