Specifications
- Antigen name:Ubiquitin specific peptidase 3
- Antigen symbol:USP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human USP3.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
USP3 is a novel human ubiquitin-specific protease. The USP3 protein is a functional ubiquitin-specific protease in vitro, and is able to inhibit ubiquitin-dependent degradation of both an N-end Rule substrate and abnormal endogenous proteins in yeast. USP3 is also only the second known ubiquitin-specific protease capable of efficiently cleaving a ubiquitin-proline bond.
USP3 antibody can be used for detection of USP3 by ELISA at 1:12500. USP3 antibody can be used for detection of USP3 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: USP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human