Anti-ZNF207 Rabbit Polyclonal Antibody
Catalog # PRSI25-147
Supplier: ProSci Inc.
Specifications
- Antigen name:Zinc Finger Protein 207
- Antigen symbol:ZNF207
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human ZNF207.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ZNF207 is a new candidate transcription factor.
ZNF207 antibody can be used for detection of ZNF207 by ELISA at 1:312500. ZNF207 antibody can be used for detection of ZNF207 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZNF207
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat