Order Entry
Spain
Orders LinkContactUsLinkComponent
Cefadroxil, Reference Standards

Cefadroxil, Reference Standards

Supplier:   USP
Cefadroxil, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1097104.
  • 1096994.
  • Description:
  • Cefadroxil
  • Cefadroxil System Suitability Mixture
  • CAS No.:
  • 66592-87-8
  • Formula:
  • C₁₆H₁₇N₃O₅S

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • Cefadroxil
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 406,00 €
    406,00 €
    1 * 200 mg
       406,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1097104.
    Description
    Cefadroxil
    CAS No.
    66592-87-8
    Formula
    C₁₆H₁₇N₃O₅S
  • 1096994.
    Cefadroxil System Suitability Mixture
    Cefadroxil System Suitability Mixture
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 370,00 €
    1 370,00 €
    1 * 25 mg
       1 370,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1096994.
    Description
    Cefadroxil System Suitability Mixture
    CAS No.
    Formula
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.