Brinzolamide, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
Specifications
- Catalog No:
- 1076363.
- 1076374.
- 1076385.
- Description:
- Brinzolamide
- Brinzolamide Related Compound A
- Brinzolamide Related Compound B
- Synonym(s):
- (S)-4-(Ethylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e] [1,2]thiazine-6-sulfonamide 1,1-dioxide), (Brinzolamide (S)-Isomer)
- (R)-4-Amino-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-6-sulfonamide 1,1-dioxide oxalate
- CAS No.:
- 138890-62-7
- 154127-19-2
- 404034-55-5
- Formula:
- C₁₂H₂₁N₃O₅S₃
- C₁₂H₂₁N₃O₅S₃
- C₁₀H₁₇N₃O₅S₃ ,C₂H₂O₄
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1076363.BrinzolamideSpecifications:
More
Specifications
Cat. No.1076363.DescriptionBrinzolamideSynonym(s)CAS No.138890-62-7FormulaC₁₂H₂₁N₃O₅S₃ - 1076374.Brinzolamide Related Compound ABrinzolamide Related Compound AUSP reference standardVial50 mgLog in for availability1 * 50 mg1 340,00 €Specifications:
More
Specifications
Cat. No.1076374.DescriptionBrinzolamide Related Compound ASynonym(s)(S)-4-(Ethylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e] [1,2]thiazine-6-sulfonamide 1,1-dioxide), (Brinzolamide (S)-Isomer)CAS No.154127-19-2FormulaC₁₂H₂₁N₃O₅S₃ - 1076385.Brinzolamide Related Compound BBrinzolamide Related Compound BUSP reference standardVial50 mgLog in for availability1 * 50 mg1 360,00 €Specifications:
More
Specifications
Cat. No.1076385.DescriptionBrinzolamide Related Compound BSynonym(s)(R)-4-Amino-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-6-sulfonamide 1,1-dioxide oxalateCAS No.404034-55-5FormulaC₁₀H₁₇N₃O₅S₃ ,C₂H₂O₄



