Order Entry
Spain
ContactUsLinkComponent
Anti-PAI1 Rabbit Polyclonal Antibody
Anti-PAI1 Rabbit Polyclonal Antibody
Catalog # BSBTA00637-2
Supplier:  Boster Bio
Anti-PAI1 Rabbit Polyclonal Antibody
Catalog # BSBTA00637-2
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PAI1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Rat,
    Mouse
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for PAI1 detection. Tested with WB, Direct ELISA in Mouse;Rat.

Type: Primary
Antigen: PAI1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat