Anti-Galectin 1 Rabbit Polyclonal Antibody
Catalog # BSBTA00470
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Galectin 1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Rat,Mouse
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Galectin 1 detection. Tested with WB, IHC-P in Mouse;Rat.
Type: Primary
Antigen: Galectin 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat