Anti-SCF Rabbit Polyclonal Antibody
BSBTA01254-1
- Antibody type:Primary
- Antigen symbol:SCF
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for SCF detection. Tested with WB, IHC-P, ELISA(Cap) in Human;Mouse;Rat.
Type: Primary
Antigen: SCF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat