Order Entry
Spain
Orders LinkContactUsLinkComponent
Baclofen, Reference Standards

Baclofen, Reference Standards

Supplier:   USP
Baclofen, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1048200.
  • 1048222.
  • Description:
  • Baclofen
  • Baclofen Related Compound A
  • Synonym(s):
  • 4-(4-Chlorophenyl)-2-pyrrolidinone
  • CAS No.:
  • 1134-47-0
  • 22518-27-0
  • Formula:
  • C₁₀H₁₂ClNO₂
  • C₁₀H₁₀ClNO
  • UN No.:
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1048200.
    Baclofen
    Baclofen
    USP reference standard
    Vial
    350 mg
    Log in for availability
    Log in for availability
    1 * 350 mg
    1 * 350 mg - 392,00 €
    392,00 €
    1 * 350 mg
       392,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1048200.
    Description
    Baclofen
    Synonym(s)
    CAS No.
    1134-47-0
    Formula
    C₁₀H₁₂ClNO₂
    UN No.
    2811
  • 1048222.
    Baclofen Related Compound A
    Baclofen Related Compound A
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 270,00 €
    1 270,00 €
    1 * 50 mg
       1 270,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1048222.
    Description
    Baclofen Related Compound A
    Synonym(s)
    4-(4-Chlorophenyl)-2-pyrrolidinone
    CAS No.
    22518-27-0
    Formula
    C₁₀H₁₀ClNO
    UN No.