Order Entry
Spain
ContactUsLinkComponent
Yttrium standard solution, NIST-3167A
Yttrium standard solution, NIST-3167A
Catalog # PROONIST-3167A
CAS Number:  
Yttrium standard solution, NIST-3167A
Catalog # PROONIST-3167A
CAS Number:  

Specifications

  • Matrix:
    10% HNO₃
  • Pk:
    5
  • Description:
    NIST-3167A
  • Element:
    Yttrium
  • Symbol:
    Y

Specifications

About this item

This standard reference material is intended for use as a primary calibration standard for the quantitative determination of yttrium.

A unit consists of five 10 ml sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically from high-purity yttrium (Ⅲ) oxide to contain a known mass fraction of yttrium.

Certifications: Traceable to NIST and SRM numbers.