Trays for Grant SUB water baths
Supplier: GRANT INSTRUMENTS
Certificates
About this item
Stainless steel perforated base trays enable baths to be used as an unstirred bath.
Frequently Bought Together
Stainless steel perforated base trays enable baths to be used as an unstirred bath.